Learn More
Medchemexpress LLC MYDGF Human His 2ug

Supplier: Medchemexpress LLC HYP705812UG
Recombinant human myeloid-derived growth factor (MYDGF), corresponding to residues S33-L173, supplied as an N-terminal 6xHis-tagged lyophilized protein expressed in E. coli. The product has a predicted molecular mass of ~18 kDa and is purified to >95% by reducing SDS-PAGE. Store frozen and reconstitute per manufacturer recommendations for research applications.
- Recombinant human protein covering residues S33-L173.
- N-terminal 6xHis tag for purification and detection.
- Expressed in E. coli and provided as a lyophilized powder.
- Predicted molecular mass ~18 kDa and >95% purity by reducing SDS-PAGE.
- Stable at -20°C; reconstitute at ≥100 μg/mL and add carrier protein for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.