Learn More
Carbosynth LLC. Myelin PLP (178-191), 1mg, MW: 1582.79, Molecular formula: C70H105N18O22S
SDP
Supplier:  Carbosynth LLC. PLP3967PI1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Myelin Proteolipid Protein (178-191). H-Asn-Thr-Trp-Thr-Thr-Cys-Gln-Ser-Ile-Ala-Phe-Pro-Ser-Lys-OH, NTWTTCQSIAFPSK, Application: Immunogenic Peptide, Storage Temperature: -20C
Catalog No. 50-168-6622