Learn More
Abcam Myelopero x idase Inhibitor, MPO inhibitor, 500MG
Supplier:  Abcam AB141482500MG
Encompass_Preferred
MW 151.17 Da, Purity >95%. Potent, irreversible and specific MPO inhibitor (IC₅₀ = 300 nM). Benzoic acid analog. No inhibition of catalase or glutathione peroxidase activity. Inhibits apoptosis in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6579