Learn More
ABclonal Technology Myeloperoxidase (MPO) Rabbit pAb
SDP
Supplier:  ABclonal Technology A1374100UL
Encompass_Preferred
Myeloperoxidase (MPO) is a heme protein synthesized during myeloid differentiation that constitutes the major component of neutrophil azurophilic granules. Produced as a single chain precursor, myeloperoxidase is subsequently cleaved into a light and heavy chain. The mature myeloperoxidase is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme produces hypohalous acids central to the microbicidal activity of neutrophils.
Catalog No. 50-000-98078