Learn More
Medchemexpress LLC MYF-01-37 | 2416417-65-5 | 99.44% | 298.30 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13960310MM/1ML
Encompass_Preferred
MYF-01-37 is a covalent TEAD inhibitor that targets Cys380 and exhibits reversible inhibition on YAP/TEAD interaction. It is intended for research use only. In vitro studies show it inhibits direct YAP/TEAD interaction in HEK 293T cells and reduces canonical YAP target gene CTGF expression in PC-9 cells, with minimal impact on cell viability in several EGFR-mutant NCSLC cell lines.
  • Covalent TEAD inhibitor
  • Targets Cys380
  • Reversibly inhibits YAP/TEAD interaction
  • Minimally impacts cell viability in EGFR-mutant NCSLC cell lines
  • Decreases dormant cells when combined with OT (osimertinib and trametinib)
Catalog No. 50-003-51968