Learn More
ABclonal Technology MYH6/α-MHC Rabbit pAb
SDP
Supplier:  ABclonal Technology A12964100UL
Encompass_Preferred
Cardiac muscle myosin is a hexamer consisting of two heavy chain subunits, two light chain subunits, and two regulatory subunits. This gene encodes the alpha heavy chain subunit of cardiac myosin. The gene is located approximately 4kb downstream of the gene encoding the beta heavy chain subunit of cardiac myosin. Mutations in this gene cause familial hypertrophic cardiomyopathy and atrial septal defect 3.
Catalog No. 50-001-14639