Learn More
Medchemexpress LLC Myo-inositol trispyrophosphate hexa-triethylamine | 851086-77-6 | 98.0% | 1213.13 | C6H12O21P6 · 6C6H15N | 5 MG
SDP
Supplier:  Medchemexpress LLC HY141510A5MG
Encompass_Preferred
myo-Inositol trispyrophosphate hexa-triethylamine is the hexa-triethylamine salt form of myo-inositol trispyrophosphate (ITPP). Supplied as a research-grade reagent (≥98.0% purity), it is used in biochemical and pharmacological studies, including investigations of oxygen delivery, tumor metabolism, and cardiovascular research. Available in small milligram pack sizes for in vitro and in vivo experiments.
  • High purity (≥98.0%) suitable for research use.
  • Supplied as a hexa-triethylamine salt to aid solubility and handling.
  • Used in studies of oxygen delivery, tumor metabolism, and cardiovascular biology.
  • Available in small milligram quantities for in vitro and in vivo applications.
  • Characterized by molecular weight 1213.13 and CAS 851086-77-6.
Catalog No. 50-002-23259