Learn More
Cayman Chemical MyoD1 RabbIt MoncLNal ntIb
SDP
Supplier:  Cayman Chemical 32294100 UL
Encompass_Preferred
A mixture of bovine ganglioside GT1b with variable fatty acyl chain lengths characterized by having two sialic residues linked to the inner galactose unit binds to the neurotoxins BTxA BTxA heavy chain and tetanus toxin (IC50s 11 0.74 and 7.2 M respectively) ganglioside GT1b-containing liposomes can bind to the major coat protein VP1 from MCPyV a putative MCPyV receptor decreases spontaneous production of IL-6 IL-10 IgG IgM and IgA in human PBMCs by 31.4 30.5 60 59.5 and 58 respectively at 10 M
Catalog No. 50-294-9195