Learn More
Genscript Corporation Myoglobin 16D3 mAb Mouse
Supplier:  Genscript Corporation V005021
Encompass
Myoglobin (symbol Mb or MB) is an iron-and oxygen-binding protein Its level in blood increases above normal levels at the beginning of myocardial necrosis associated with myocardial infarction (MI) It is used as a diagnosis marker for skeletal or myocardial muscle injury
Catalog No. 50-000-58740