Learn More
Medchemexpress LLC Myoseverin | 267402-71-1 | 99.4% | 432.52 | 100 MG
Supplier: Medchemexpress LLC HYW008956100MG
Myoseverin is a microtubule-binding molecule and an angiogenesis inhibitor. It can induce the reversible fission of multinucleated myotubes into mononucleated fragments. Additionally, Myoseverin demonstrates anti-angiogenic effects by inhibiting endothelial cell function and the differentiation of endothelial progenitor cells.
- Microtubule-binding molecule
- Angiogenesis inhibitor
- Induces reversible fission of multinucleated myotubes into mononucleated fragments
- Exerts anti-angiogenic effects by inhibiting endothelial cell function
- Inhibits endothelial progenitor cell differentiation