Learn More
Medchemexpress LLC Myosmine | 532-12-7 | 146.19 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00190910MM/1ML
Encompass_Preferred
Myosmine is a specific tobacco alkaloid found in nuts and nut products. It acts as an nAChR activator and has a low affinity for a4b2 nicotinic acetylcholinergic receptors (nAChR) with a Ki of 3300 nM. It is easily nitrosated, yielding 4-hydroxy-1-(3-pyridyl)-1-butanone (HPB) and the esophageal tobacco carcinogen N'-nitrosonornicotine. This product is for research use only.
  • Acts as an nAChR activator
  • Low affinity for a4b2 nicotinic acetylcholinergic receptors
  • Easily nitrosated to form carcinogens
  • For research use only
Catalog No. 50-004-46088