Learn More
Sigma Aldrich Fine Chemicals Biosciences MYPT1 654 880
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 12457
Encompass
ROK substrate protein corresponding to amino acids 654-880 of human MYPT1 with an N-terminal GST tag. MW 60 kDa.
Catalog No. 50-290-5734