Learn More
Cayman Chemical Myricetin, 529-44-2, 10 mg
SDP
Supplier:  Cayman Chemical 1001260010
Encompass_Preferred
Molecular Formula C15H10O8, Purity ≥98%, Formula Weight 318.2, Synonyms: Cannabiscetin, NSC 407290; A flavonoid compound that acts as a powerful antioxidant; inhibits TBARS formation with an IC50 value of 6.34 M; blocks oxLDL uptake by U937-derived macrophages at 20 M; demonstrates potent chemopreventative potential by binding JAK1/STAT3 to inhibit neoplastic transformation of murine JB6 Pt cells.
Catalog No. NC0480358