Learn More
Cayman Chemical MyrIstIc AcId-d27 250mg
SDP
Supplier:  Cayman Chemical 9003317250 MG
Encompass_Preferred
An internal standard for the quantification of myristic acid by GC- or LC-MS
Catalog No. 50-289-4245