Learn More
Medchemexpress LLC Myristic acid-d5 | 327077-03-2 | 233.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN2041S61MG
Encompass_Preferred
Myristic acid-d5 is the deuterium labeled Myristic acid, which is a saturated 14-carbon fatty acid found in most animal and vegetable fats, including butterfat and coconut, palm, and nutmeg oils.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15900