Learn More
Medchemexpress LLC Myristic anhydride 100g
SDP
Supplier:  Medchemexpress LLC HYW127537100G
Encompass_Preferred
Myristic anhydrideIt is an organic compound belonging to acid anhydrides It is formed by the reaction of myristic acid and acetic anhydride Myristic anhydrideIt has various applications in the chemical and pharmaceutical industries especially as an intermediate in the synthesis of various chemicals and pharmaceuticals In addition it can also be used as a curing agent for epoxy resins
Catalog No. 50-002-06326