Learn More
Medchemexpress LLC Myristoyl glutamic acid sodium | 38517-37-2 | 98.4% | 10 G
Supplier: Medchemexpress LLC HY126460A10G
Myristoyl glutamic acid sodium is a common ingredient found in cosmetics/creams. It is a surfactant belonging to the amino acid family.
- Possesses mild cleaning properties.
- Foamy.
- Mildly bacteriostatic.