Learn More
Cayman Chemical MyrIstoyl GlycIn 5mg
SDP
Supplier:  Cayman Chemical 301075 MG
Encompass_Preferred
A lipidated amino acid; induces browning of SGBS preadipocytes from 0.0001-20 µM; has been used as an immunogen to raise antibodies against myristoylated proteins; a synthetic intermediate in the synthesis of lipopeptides
Catalog No. 50-299-0509