Learn More
Medchemexpress LLC Myristoyl pentapeptide-4 | 1392416-25-9 | 98.9% | 774.00 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP523910MG
Encompass_Preferred
Myristoyl pentapeptide-4 is a bioactive peptide known for its hair growth promotion effects and is used as a cosmetic ingredient. This synthetic peptide (KTTKS) targets keratin, activating keratin gene transcription and translation to improve hair elasticity and strength. It also promotes the proliferation of HaCaT cells, stimulates the migration of human hair follicle papilla cells (HDPCs), and encourages skin fibroblasts to produce collagen and fibronectin, which can reduce wrinkles, improve skin elasticity, and aid in wound healing and skin repair.
  • Promotes hair growth
  • Targets keratin to improve hair elasticity and strength
  • Promotes proliferation of HaCaT cells
  • Stimulates migration of human hair follicle papilla cells (HDPCs)
  • Stimulates skin fibroblasts to produce collagen and fibronectin
  • Reduces wrinkles and improves skin elasticity
  • Promotes wound healing and skin repair
Catalog No. 50-003-99186