Learn More
Medchemexpress LLC Myristoyl pentapeptide-4 | 1392416-25-9 | 98.9% | 774.00 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP52395MG
Encompass_Preferred
Myristoyl pentapeptide-4 is a bioactive synthetic peptide composed of five amino acids (Lys-Thr-Thr-Lys-Ser). It is used as a cosmetic ingredient, promoting hair growth by targeting keratin and activating gene transcription and translation to improve hair elasticity and strength. It also aids in skin health by stimulating collagen and fibronectin production, reducing wrinkles, and promoting wound healing and skin repair.
  • Promotes hair growth
  • Improves hair elasticity and strength
  • Promotes proliferation of HaCaT cells
  • Stimulates migration of human hair follicle papilla cells (HDPCs)
  • Stimulates skin fibroblasts to produce collagen and fibronectin
  • Reduces wrinkles and improves skin elasticity
  • Promotes wound healing and skin repair
Catalog No. 50-003-99726