Learn More
Carbosynth LLC. Myr-Ser-Ile-Tyr-Arg-Arg-Gly-Ala-Arg-Arg-Trp-Arg-Lys-Leu, 1mg, MW: 1928.4, Molecular formula: C90H154N30O17
SDP
Supplier:  Carbosynth LLC. PKC3403V1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Myristoyl-Ser-Ile-Tyr-Arg-Arg-Gly-Ala-Arg-Arg-Trp-Arg-Lys-Leu[Myristoyl-PKC Zeta (113-125) TFA, Application: Myristoylated Cell Permeable PKC Pseudosubstrate Inhibitor, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20C, References: C. Laudanna, D. Mochly-Rosen, T. Liron, G. Constantin, and E.C. Butcher, J. Biol. Chem., 273, 30306 (1998). (Pharmacol.), J. Biol. Chem., 273, 30306 (1998). (Pharmacol.), S. Lim, J.W. Choi, H.S. Kim, Y.-H. Kim, K. Yea, K. Heo, J.H. Kim, S.-H. Kim, M. Song, J.I. Kim, S.H. Ryu, and P.-G. Suh, Life Sci., 82, 733 (2008). (Biochem.)
Catalog No. 50-168-6586