Learn More
Medchemexpress LLC (-)-Myrtenal | 18486-69-6 | 99.1% | 150.22 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY121401A500MG
Encompass_Preferred
(-)-Myrtenal is an orally active terpene exhibiting antitumour activity. It ameliorates hyperglycemia by enhancing GLUT2 through Akt in the skeletal muscle and liver of diabetic rats. In vitro studies demonstrate a strong cytotoxic effect on human colon tumour (HT29) and normal colon epithelial cells (CCD 841 CoTr).
  • Orally active terpene
  • Exhibits antitumour activity
  • Ameliorates hyperglycemia by enhancing GLUT2 through Akt
  • Strong cytotoxic effect on human colon tumour and normal colon epithelial cells
  • Decreases plasma glucose levels in diabetic rats
  • Improves plasma insulin levels in diabetic rats
  • Up-regulates IRS2, Akt, and GLUT2 in liver
  • Up-regulates IRS2, Akt, and GLUT4 protein expression in skeletal muscle
  • Appearance: liquid
  • Color: colorless to light yellow
  • Initial source: plants (Leguminosae Erythrina poeppigiana, Labiatae)
Catalog No. 50-004-37085