Learn More
Selleck Chemical LLC MZ-1 S8889-100MG
SDP
Supplier:  Selleck Chemical LLC S8889100MG
Encompass_Preferred
MZ-1 is a PROTAC degrader of bromodomain-containing protein 4 (BRD4) MZ-1 binds to the Brd bromodomain with Kd of 62 nM 60 nM 21 nM 13 nM 39 nM and 15 nM for Brd2BD1 Brd2BD2 Brd3BD1 Brd3BD2 Brd4BD1 and Brd4BD2
Catalog No. 50-000-24705