Learn More
Medchemexpress LLC MZP-54 | 2010159-47-2 | 98.0% | 1036.67 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11237610MG
Encompass_Preferred
MZP-54 is a PROTAC connected by ligands for von Hippel-Lindau and BRD3/4, exhibiting a Kd of 4 nM for Brd4BD2. This selective degrader is based on PROTAC technology, binding to VHL-EloC-EloB protein (VCB) with a Kd of 105 ± 24 nM. It shows inhibitory activity against MV4;11 and HL60 cells, and exhibits high depletion of cMyc levels.
  • Selective degrader of BRD3/4
  • Based on PROTAC technology
  • Binds to VHL-EloC-EloB protein (VCB)
  • Shows inhibitory activity against MV4;11 and HL60 cells
  • Exhibits high depletion of cMyc levels
Catalog No. 50-196-9086