Learn More
Medchemexpress LLC N-[(1,1-Dimethylethoxy)carbonyl]-D-2,3-diaminopropanoic acid | 76387-70-7 | MFCD01632072 | ≥97.0% | 204.22 g/mol | C8H16N2O4 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW014692100G
Encompass_Preferred
Boc-D-2,3-diaminopropionic acid (N-t-Boc-amino-D-alanine) is a Boc-protected amino acid derivative commonly used as a building block in peptide synthesis and medicinal chemistry research. The compound is the D-stereoisomer of 2,3-diaminopropionic acid and is supplied as a white to off-white solid with high purity and established storage recommendations.
  • Boc protection on the amino group facilitates selective coupling and deprotection.
  • D-stereoisomer suitable for chiral-specific peptide assembly.
  • High purity (≥97%) supports reproducible synthetic results.
  • Solid form with defined storage stability for long-term use.
  • Available in multiple laboratory-scale package sizes, including 100 g.
Catalog No. 50-003-02631