Learn More
Medchemexpress LLC N-((1,1-Dimethylethoxy)carbonyl)-L-alanyl-L-alanine | 27317-69-7 | MFCD00038564 | 99.0% | 260.29 g/mol | C11H20N2O5 | 10 G

Supplier: Medchemexpress LLC HYW03975610G
Boc-Ala-Ala-OH (NSC 334362) is a Boc-protected L-alanyl-L-alanine dipeptide used as a research reagent and intermediate in the synthesis of antibacterial agents. It is typically handled in laboratory peptide synthesis and medicinal chemistry workflows and is supplied for research use only.
- Boc-protected dipeptide useful as a peptide synthesis intermediate.
- Commonly used in preparation of antibacterial agents.
- Chemical formula C11H20N2O5.
- Molecular weight 260.29 g/mol.
- CAS 27317-69-7.
- Typical purity 99.04%.
- Available in multiple lab-scale pack sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.