Learn More
Cayman Chemical N-1- 2 3-DIoleyloxypropyl- 5mg
SDP
Supplier:  Cayman Chemical 259265 MG
Encompass_Preferred
A cationic lipid; has been used as a component in liposomes that can be used to encapsulate siRNA, microRNAs, and oligonucleotides and for gene transfection in vitro
Catalog No. 50-293-3360