Learn More
Medchemexpress LLC N'-(1-(3-bromophenyl)ethylidene)-2-oxo-4-phenylpyrrolidine-3-carbohydrazide | 1051487-82-1 | MFCD18086869 | 98.3% | 400.27 g·mol⁻¹ | C19H18BrN3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY143515MG
Encompass_Preferred
AC-264613 is a potent, selective small-molecule agonist of protease-activated receptor 2 (PAR-2), reported with pEC50 ≈ 7.5 and provided as a research reagent.
  • Potent protease-activated receptor 2 (PAR-2) agonist (pEC50 ≈ 7.5).
  • Selective activity for PAR-2 over related targets.
  • High purity: 98.3% (HPLC reported).
  • Molecular formula C19H18BrN3O2; molecular weight 400.27 g·mol⁻¹.
  • Provided as a 5 MG research reagent for laboratory use.
Catalog No. 50-002-22683