Learn More
Medchemexpress LLC N-(1-acetyl-2,3-dihydroindol-6-yl)-3-(3-cyanophenyl)-N-(1-(2-cyclopentylethyl)piperidin-4-yl)prop-2-enamide | 683746-68-1 | MFCD19690929 | 98.0% | 510.67 | C32H38N4O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1077321MG
Encompass_Preferred
This compound is a nonpeptidic antagonist of the neuropeptide Y Y2 receptor used as a research tool for pharmacology and signaling studies. It offers submicromolar potency at human and rodent receptors and high subtype selectivity.
  • Nonpeptidic antagonist of the neuropeptide Y Y2 receptor.
  • Submicromolar potency (pIC50 ≈ 7.0 human, 7.1 rat).
  • Greater than 100-fold selectivity versus Y1, Y4, and Y5 receptors.
  • Powder form, soluble in DMSO (≈5 mg/mL) with warming and sonication.
  • Stable as powder at -20°C for long-term storage; in solvent store frozen.
  • Molecular weight 510.67; molecular formula C32H38N4O2.
Catalog No. 50-002-41218