Learn More
Medchemexpress LLC N-(1-benzyl-2,3-dihydropyrrolo[2,3-b]quinolin-4-yl)-2-phenylacetamide | 365565-02-2 | 99.3% | 500 UG
SDP
Supplier:  Medchemexpress LLC HY119823500UG
Encompass_Preferred
PGP-4008 is a research-grade small molecule P-glycoprotein (Pgp) inhibitor used in preclinical and in vitro studies. It is supplied as a solid with high purity and has been reported to inhibit tumor growth in murine Pgp-mediated multiple agent resistance models when combined with doxorubicin.
  • Specific P-glycoprotein inhibitor for multidrug resistance research
  • High purity suitable for biochemical and cell-based assays
  • Available in small-scale quantities and solution formats for flexible use
  • Soluble in DMSO at 62.5 mg/mL for convenient stock preparation
  • Stable as a powder at -20°C for extended shelf life
Catalog No. 50-002-31874