Learn More
Cayman Chemical N 1-Carboxyethyl-L-lysIn 5mg
SDP
Supplier:  Cayman Chemical 253335 MG
Encompass_Preferred
An advanced glycation end product; protein bound, but not free CEL, binds to RAGE; elevated in the lens of diabetic patients with cataract; decreases glutamate uptake and secretion of S100B in rat hippocampal slices in a RAGE-independent manner at 1 mM
Catalog No. 50-293-2471