Learn More
Medchemexpress LLC N-(1-ethylpropyl)-3,6-dimethyl-2-(2,4,6-trimethylphenoxy)-4-pyridinamine | 175140-00-8 | MFCD14584860 | >98.0% | C21H30N2O | 10MG
SDP
Supplier:  Medchemexpress LLC HY1413010MG
Encompass_Preferred
CP 376395 is a potent, selective, brain-penetrant corticotropin-releasing factor 1 (CRF1) receptor antagonist used in preclinical pharmacology and neuroscience research. It is supplied as a research-grade solid with documented identity and analytical characterization.
  • Potent, selective CRF1 receptor antagonist useful for mechanistic studies.
  • Brain-penetrant for in vivo central nervous system research.
  • High purity suitable for biological assays (typically >98.0% by HPLC).
  • Characterized by molecular formula C21H30N2O and molecular weight 326.48 g/mol.
  • Provided with analytical documentation such as certificate of analysis and datasheet.
Catalog No. 50-002-59309