Learn More
Enzo Life Sciences N-(1-Naphthalenylsulfonyl)-Ile-Trp-aldehyde (1mg). CAS: 161709-56-4
SDP
Supplier:  Enzo Life Sciences ALX260133M001
Encompass_Preferred
Cathepsin-L inhibitor. Potent, selective and reversible inhibitor of human cathepsin L (IC50=1.9nM). Inhibits release of Ca2+ and hydroxyproline from bone in vitro. Alternative name: N-(1Naphthalenylsulfonyl)- IW-CHO. Formula: C27H29N3O4S. MW: 491.6. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO or ethyl acetate (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8336