Learn More
Medchemexpress LLC N-[(1R)-1-(3-amino-5-(trifluoromethyl)phenyl)ethyl]-7-methoxy-2-methylquinazolin-4-amine | 2230836-55-0 | MFCD32197204 | 100.0% | 462.46 g/mol | C23H25F3N4O3 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12581725MG
Encompass_Preferred
BI-3406 is a potent, selective, orally active small-molecule inhibitor of the SOS1-KRAS protein-protein interaction used as a research tool in oncology. It reduces formation of GTP-loaded RAS and limits proliferation in KRAS-driven cancer models; supplied as a white to off-white solid for laboratory use.
  • Potent SOS1-KRAS interaction inhibitor with low-nanomolar activity.
  • Suitable for biochemical and cellular assays.
  • High chemical purity for research applications.
  • Supplied as a white to off-white solid sample for laboratory use.
  • Soluble in common organic solvents such as DMSO for assay preparation.
Catalog No. 50-002-36572