Learn More
Medchemexpress LLC N-[(1R)-2,3-dihydro-1H-inden-1-yl]-adenosine | 96392-15-3 | MFCD00876438 | 99.97% | 383.40 g/mol | C19H21N5O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10003225MG
Encompass_Preferred
PD 117519 (CI-947) is a synthetic A2A adenosine receptor agonist used as a research reagent in pharmacological studies. It has demonstrated oral antihypertensive activity in animal models and is provided as a high-purity sample for both in vitro and in vivo experiments.
  • High purity suitable for analytical and pharmacological studies.
  • Well characterized chemical identity with supporting safety and product information.
  • Soluble in DMSO for straightforward preparation of stock solutions.
  • Appropriate for A2A receptor pharmacology and antihypertensive mechanism research.
  • Provided in mg-scale packaging for small-scale laboratory studies.
Catalog No. 50-002-15066