Learn More
Medchemexpress LLC N-[(1S)-2-([3-bromo-4,5-dihydro-5-isoxazolyl]methylamino)-1-(4-hydroxyphenyl)methyl-2-oxoethyl] carbamic acid, benzyl ester | 744198-19-4 | ≥98.0% | 476.3 | C21H22BrN3O5 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12329025MG
Encompass_Preferred
KCC009 is a small-molecule inhibitor of transglutaminase 2 (TG2) used as a research reagent; it has been reported to induce p53-independent radiosensitization and to disrupt fibronectin assembly in the extracellular matrix. Molecular formula C21H22BrN3O5, formula weight 476.3, purity ≥98%.
  • TG2 inhibition for mechanistic studies.
  • Reported to induce p53-independent radiosensitization in cellular models.
  • Disrupts fibronectin assembly in the extracellular matrix.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C21H22BrN3O5; formula weight 476.3.
Catalog No. 50-002-37376