Learn More
Medchemexpress LLC N-[(1S)-3-[(2Z)-2-[(4R)-3,4-dimethyl-1,3-thiazolidin-2-ylidene]hydrazinyl]-1-(oxan-4-yl)-2,3- | 868273-90-9 | 99.6% | 438.58 g·mol⁻¹ | C21H34N4O4S | 100 MG

Supplier: Medchemexpress LLC HY108044100MG
Small-molecule research compound originally developed as a cathepsin K inhibitor for the treatment of osteoporosis; now used in preclinical studies to probe cathepsin K biology and investigate antiviral activity. Supplied as a solid powder for in vitro and animal research use only, not for human therapeutic use.
- Potent, selective cathepsin K inhibitor.
- Intended for in vitro and preclinical research.
- Solid powder form, off-white to light yellow.
- High purity (≈99.6%), batch-dependent.
- Molecular formula C21H34N4O4S; molecular weight 438.58 g·mol⁻¹.
- CAS registry number 868273-90-9.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.