Learn More
Medchemexpress LLC N-[2-(1,3-benzodioxol-5-yl)ethyl]-1-(2-imidazol-1-yl-6-methylpyrimidin-4-yl)pyrrolidine-2-carboxamide | 402934-09-2 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1212410MM 1ML
Encompass_Preferred
BBS-4 is a potent, selective inducible nitric oxide synthase (iNOS/NOS2) dimerization inhibitor with a reported IC50 of 0.49 nM. It has been shown to protect mice from sepsis-associated cardiovascular dysfunction. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL vial) and is intended for preclinical research applications.
  • Potent iNOS dimerization inhibitor with IC50 0.49 nM.
  • Ready-to-use 10 mM solution in DMSO, supplied in a 1 mL vial.
  • High purity (99.51% by HPLC) suitable for research use.
  • Molecular weight 420.46; molecular formula C22H24N6O3.
  • Intended for in vitro and in vivo preclinical studies.
Catalog No. 50-002-31765