Learn More
Medchemexpress LLC N-[2-(2-cyclopropylmethoxyphenoxy)ethyl]-5-chloro-α,α-dimethyl-1H-indole-3-ethanamine HCl | 169505-93-5 | 99.9% | 449.41 | C24H30Cl2N2O2 | 1 ML

Supplier: Medchemexpress LLC HY10133610MM 1ML
A selective α1A adrenoceptor antagonist supplied as a ready-to-use 10 mM solution in DMSO for in vitro pharmacology and receptor signaling studies. Demonstrates high potency and is provided at high purity for research applications.
- Supplied as a 10 mM solution in DMSO, 1 mL.
- High potency: pKi 9.1 and pA2 9.8.
- High purity: 99.9%.
- Molecular weight: 449.41.
- CAS number: 169505-93-5.
- Storage: store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.