Learn More
Medchemexpress LLC N-(2-(4-(3,5-dioxo-1,2,4-triazolidin-4-yl)-2-methoxyphenoxy)ethyl)-4,7,10,13-tetraoxahexadec-1-5-ynamide | 99.3% | 508.52 g/mol | C23H32N4O9 | 5 MG

Supplier: Medchemexpress LLC HY1360465MG
PTAD-PEG4-alkyne is a cleavable four-unit polyethylene glycol (PEG) linker bearing an alkyne group for click chemistry and antibody-drug conjugate synthesis. It enables conjugation to azide-functionalized molecules via copper-catalyzed azide-alkyne cycloaddition and provides a hydrophilic, cleavable spacer for payload attachment.
- Contains terminal alkyne for CuAAC (click) conjugation.
- Four-unit PEG spacer provides hydrophilicity and cleavability for ADC applications.
- High purity (99.3%) and molecular weight approximately 508.52 g/mol.
- Available in small research pack sizes, for example 5 mg.
- Storage: powder at -20°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.