Learn More
Medchemexpress LLC N-((2,4-dimethylphenyl)sulfonyl)-2-(naphthalen-2-yloxy)acetamide | 868145-09-9 | 99.4% | 369.43 g/mol | C20H19NO4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1297385MG
Encompass_Preferred
I942 is a selective non-cyclic nucleotide (NCN) EPAC1 agonist used as a research tool to modulate EPAC1-dependent signaling. It attenuates proinflammatory cytokine signaling associated with cardiovascular disease and is supplied as a high-purity reagent for biochemical and cellular studies.
  • Selective EPAC1 agonist useful for signaling studies.
  • Non-cyclic nucleotide scaffold enables cellular EPAC1 activation.
  • High purity suitable for biochemical and cellular assays.
  • Molecular weight 369.43 g/mol; molecular formula C20H19NO4S.
  • Supplied in small milligram quantities for research use.
  • Relevant to GPCR/G protein and MAPK/ERK pathway investigations.
Catalog No. 50-002-62082