Learn More
Medchemexpress LLC N-[2-(4-oxo-1-phenyl-1,3,8-triazaspirodec-8-yl)ethyl]-2-naphthalenecarboxamide | 1130067-34-3 | >98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY121983100MG
Encompass_Preferred
This small-molecule compound (CAS 1130067-34-3) is a potent phospholipase D2 (PLD2) inhibitor used for biochemical and cell-based research. It has reported in vitro IC50 = 140 nM and cellular IC50 = 110 nM, and is supplied at high purity for research applications.
  • Potent PLD2 inhibitor with reported IC50 values of 140 nM (in vitro) and 110 nM (cellular).
  • High purity (>98.0%).
  • Available as solid in multiple pack sizes and as a 10 mM solution in DMSO.
  • Suitable for in vitro biochemical assays and cell-based studies of migration and signaling.
  • Molecular formula C26H28N4O2; molecular weight 428.5 g/mol.
Catalog No. 50-002-31606