Learn More
Medchemexpress LLC N-[2,5-bis(trifluoromethyl)phenyl]-5-bromo-2-hydroxybenzamide | 439144-66-8 | 98.7% | 428.12 g·mol⁻¹ | C15H8BrF6NO2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10566125MG
Encompass_Preferred
IMD-0560 is a small-molecule inhibitor of IκB kinase β (IKKβ) supplied for research use. The compound is provided as a solid with high analytical purity and is used to probe NF-κB signaling in biochemical and cellular assays. Not for human or clinical use.
  • Selective IκB kinase β inhibition for pathway-focused studies.
  • High purity (98.7%) for reproducible experimental results.
  • Solid form compatible with common solvent dissolution protocols.
  • Molecular weight 428.12 g·mol⁻¹ and formula C15H8BrF6NO2 for precise dosing.
  • Available in small research quantities, suitable for pilot and in vitro work.
  • Documentation available including data sheet, COA, and SDS.
Catalog No. 50-002-40914