Learn More
Medchemexpress LLC N-[2,6-dibromo-4-(1-methylethyl)phenyl]-N'-3-pyridinyl-urea | 688309-70-8 | MFCD02678819 | 99.2% | 413.11 | C15H15Br2N3O | 5 MG

Supplier: Medchemexpress LLC HY1102375MG
BX430 is a potent, selective, noncompetitive allosteric antagonist of human P2X4 receptor channels used as a research tool in studies of P2X4-mediated signaling, chronic pain, and cardiovascular disease. It is provided as a high-purity solid with recommended storage and solubility guidance for in vitro and in vivo use.
- Potent, selective noncompetitive allosteric antagonist of P2X4 receptors.
- CAS number: 688309-70-8.
- High purity: 99.21% by HPLC.
- Molecular weight 413.11 and formula C15H15Br2N3O.
- Soluble in DMSO (≈83.33 mg/mL); reported in vivo formulations available.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.