Learn More
Medchemexpress LLC N-(2-amino-5-(thiophen-2-yl)phenyl)-2-(piperazin-1-yl)quinoline-6-carboxamide | 1609389-52-7 | MFCD30342405 | 99.6% | 429.54 g·mol⁻¹ | C24H23N5OS | 25 MG
SDP
Catalog No. 5000243614
Encompass_Preferred
ACY-957 is a small-molecule, orally active inhibitor selective for histone deacetylases HDAC1 and HDAC2 used in preclinical research. It potently inhibits HDAC1 (IC50 = 7 nM) and HDAC2 (IC50 = 18 nM), with substantially weaker activity against HDAC3 (IC50 = 1,300 nM), and shows no significant inhibition of HDAC4-HDAC9. The compound is provided as a purified research reagent for biochemical and cellular studies of epigenetic regulation.
  • Potent HDAC1 inhibition (IC50 = 7 nM).
  • Selective for HDAC1/2 over HDAC3 (IC50 HDAC2 = 18 nM; HDAC3 = 1,300 nM).
  • Orally active profile reported in preclinical studies.
  • Suitable for biochemical and cellular assays investigating epigenetic regulation.
  • High chemical purity for reproducible experimental results.
Catalog No. 50-002-43614 Supplier Medchemexpress LLC Supplier No. HY10400825MG
ACY-957 is a small-molecule, orally active inhibitor selective for histone deacetylases HDAC1 and HDAC2 used in preclinical research. It potently inhibits HDAC1 (IC50 = 7 nM) and HDAC2 (IC50 = 18 nM), with substantially weaker activity against HDAC3 (IC50 = 1,300 nM), and shows no significant inhibition of HDAC4-HDAC9. The compound is provided as a purified research reagent for biochemical and cellular studies of epigenetic regulation.
  • Potent HDAC1 inhibition (IC50 = 7 nM).
  • Selective for HDAC1/2 over HDAC3 (IC50 HDAC2 = 18 nM; HDAC3 = 1,300 nM).
  • Orally active profile reported in preclinical studies.
  • Suitable for biochemical and cellular assays investigating epigenetic regulation.
  • High chemical purity for reproducible experimental results.