Learn More
Medchemexpress LLC N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide | 36321-43-4 | MFCD08529471 | 95.0% | 228.29 g·mol⁻¹ | C14H16N2O | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW140566100MG
Encompass_Preferred
N-(2-aminoethyl)-2-(naphthalen-1-yl)acetamide is an analytical standard and an identified impurity of naphazoline used as a drug intermediate or reference material in analytical and synthetic workflows. Molecular formula C14H16N2O, molecular weight 228.29 g·mol⁻¹, CAS 36321-43-4. Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Suitable as an analytical standard for impurity profiling.
  • Intended for method development and validation.
  • Provided in a 100 MG quantity for laboratory use.
  • Stable under recommended storage; protect from light.
  • Applicable as a drug intermediate or reference material.
Catalog No. 50-002-83576