Learn More
Matrix Scientific N-(2-Azepan-1-yl-phenyl)-2-chloro-acetamide, 436087-22-8, MFCD03446301, 1g
Supplier: Matrix Scientific 0085851G
Molecular Formula C14H19ClN2O, Molecular Weight 266.77
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.