Learn More
Medchemexpress LLC N-[2-chloro-5-(trifluoromethyl)phenyl]-2-[4-(furan-2-carbonyl)piperazin-1-yl]acetamide | 899713-86-1 | MFCD05123530 | 99.0% | 415.79 g/mol | C18H17ClF3N3O3 | 25 MG

Supplier: Medchemexpress LLC HY10073625MG
ML348 (GNF-Pf-1127) is a selective, reversible inhibitor of acyl-protein thioesterase 1 (APT1/LYPLA1) used as a research tool to probe depalmitoylation and lipid signaling pathways. It is supplied as a solid with documented solubility and storage conditions for in vitro and in vivo studies.
- Selective and reversible APT1 (LYPLA1) inhibitor (IC50 ≈ 210 nM).
- Minimal activity against LYPLA2, enabling target-specific studies.
- Soluble in DMSO (25 mg/mL) for in vitro use; in vivo formulation ≥ 2.5 mg/mL in 10% DMSO/90% corn oil.
- Powder stability: -20 °C for up to 3 years; in solvent stability: -80 °C recommended.
- Molecular weight 415.79 g/mol and chemical formula C18H17ClF3N3O3 for experimental calculations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.