Learn More
Medchemexpress LLC N-(2-hydroxyethyl)iminodiacetic acid | 93-62-9 | MFCD00004293 | 97.0% | 177.16 | C6H11NO5 | 25 G
SDP
Supplier:  Medchemexpress LLC HYW12778325G
Encompass_Preferred
N-(2-Hydroxyethyl)iminodiacetic acid is an amino-acid derivative used primarily as a metal-chelating reagent in research applications. Supplied as a research-use chemical (CAS 93-62-9) with a reported purity of 97.0% and molecular weight 177.16 g/mol, it is provided in laboratory pack sizes for analytical and synthetic workflows.
  • Chelates metal ions for analytical and preparative applications.
  • Suitable for research use in synthesis and method development.
  • Provided at 97.0% purity for consistent experimental results.
  • Available in laboratory pack sizes for convenient handling.
Catalog No. 50-002-83623