Learn More
Medchemexpress LLC N-(2-hydroxyethyl)iminodiacetic acid | 93-62-9 | MFCD00004293 | >=98.0% | 177.16 g/mol | C6H11NO5 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW127783100G
Encompass_Preferred
N-(2-Hydroxyethyl)iminodiacetic acid is an amino acid derivative used as a chelating and buffering reagent in laboratory research. It is supplied as a powder for biochemical and analytical applications where metal ion complexation or buffer formulation is required. Identifiers include CAS 93-62-9, formula C6H11NO5, and molecular weight 177.16 g/mol.
  • Chelating agent for metal ions including Fe(III)
  • Useful as a buffer component in biochemical assays
  • Solid powder form convenient for weighing and storage
  • Available in multiple pack sizes for laboratory use
  • High purity suitable for research (>=98.0%)
  • For research use only; not for human or clinical applications
Catalog No. 50-002-83513